Learn More
Research Products International Corp DL-PHOSPHINOTHRICIN [PPT], 250 MG
SDP
Catalog No. 50213617
Encompass_Preferred
Analog of Glutamate that acts as a competitive inhibitor of Glutamine synthetase. This enzyme is involved in assimilation of ammonia and plays a key role in nitrogen assimilation.
Catalog No. 50-213-617 Supplier Research Products International Corp Supplier No. P401000.25
Analog of Glutamate that acts as a competitive inhibitor of Glutamine synthetase. This enzyme is involved in assimilation of ammonia and plays a key role in nitrogen assimilation.