Learn More
Medchemexpress LLC DL-Proline-d3 | 784086-28-8 | 99.7% | 118.15 | 10 MG
SDP
Catalog No. 5000366748
Encompass_Preferred
DL-Proline-d3 is a deuterium labeled amino acid, appearing as a white to off-white solid. It is intended for research use only in laboratory settings and for the manufacture of substances. This product has been tested and complies with its given specifications, but it has not been fully validated for medical applications.
  • High purity (99.67% by HPLC)
  • High isotopic enrichment (99.4%)
  • Suitable for use as laboratory chemicals
  • Suitable for the manufacture of substances
  • White to off-white solid appearance
Catalog No. 50-003-66748 Supplier Medchemexpress LLC Supplier No. HYW012908S210MG
DL-Proline-d3 is a deuterium labeled amino acid, appearing as a white to off-white solid. It is intended for research use only in laboratory settings and for the manufacture of substances. This product has been tested and complies with its given specifications, but it has not been fully validated for medical applications.
  • High purity (99.67% by HPLC)
  • High isotopic enrichment (99.4%)
  • Suitable for use as laboratory chemicals
  • Suitable for the manufacture of substances
  • White to off-white solid appearance