Learn More
Medchemexpress LLC DL-propargylglycine | 50428-03-0 | MFCD00056728 | 98.0% | 113.11 | C5H7NO2 | 500 MG
SDP
Catalog No. 5000299524
Encompass_Preferred
DL-Propargylglycine is an alkyne-bearing amino acid derivative used as a reagent in click chemistry and synthetic applications. It provides a terminal alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is supplied for research use with supporting purity and safety documentation.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for bioconjugation and labeling in chemical biology workflows.
  • Amino acid derivative useful as a synthetic building block.
  • High purity (98.0%) with certificate of analysis and safety data sheet available.
  • Offered in research-scale quantities (100 mg-1 g) with larger amounts available by quote.
Catalog No. 50-002-99524 Supplier Medchemexpress LLC Supplier No. HYW040124500MG
DL-Propargylglycine is an alkyne-bearing amino acid derivative used as a reagent in click chemistry and synthetic applications. It provides a terminal alkyne handle for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and is supplied for research use with supporting purity and safety documentation.
  • Contains a terminal alkyne for copper-catalyzed azide-alkyne cycloaddition (CuAAC).
  • Suitable for bioconjugation and labeling in chemical biology workflows.
  • Amino acid derivative useful as a synthetic building block.
  • High purity (98.0%) with certificate of analysis and safety data sheet available.
  • Offered in research-scale quantities (100 mg-1 g) with larger amounts available by quote.