Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-propargylglycine | 50428-03-0 | MFCD00056728 | 98.0% | 113.11 g/mol | C5H7NO2 | 1 G
SDP

Catalog No. 5000299472
Encompass_Preferred

DL-propargylglycine is a small-molecule amino acid derivative bearing a terminal alkyne, commonly used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is provided as a research-grade compound with high purity suitable for synthetic chemistry and chemical biology workflows such as conjugation, labeling, and method development.

  • Contains terminal alkyne functionality for CuAAC reactions.
  • High purity suitable for research applications.
  • Racemic (DL) amino acid derivative.
  • Simple molecular composition (C5H7NO2; 113.11 g/mol).
  • Store at 4°C and protect from light for optimal stability.

Catalog No. 50-002-99472 Supplier Medchemexpress LLC Supplier No. HYW0401241G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-propargylglycine is a small-molecule amino acid derivative bearing a terminal alkyne, commonly used as a click-chemistry reagent for copper-catalyzed azide-alkyne cycloaddition (CuAAC). It is provided as a research-grade compound with high purity suitable for synthetic chemistry and chemical biology workflows such as conjugation, labeling, and method development.

  • Contains terminal alkyne functionality for CuAAC reactions.
  • High purity suitable for research applications.
  • Racemic (DL) amino acid derivative.
  • Simple molecular composition (C5H7NO2; 113.11 g/mol).
  • Store at 4°C and protect from light for optimal stability.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.