Learn More
Cayman Chemical DL-Pyroglutamic Acid, Purity: ≥95%, CAS: 149-87-1, 500 g
SDP
Catalog No. 502912053
Encompass_Preferred
A cyclized form of glutamic acid; decreases immobility time in the forced swim test and increases the latency to paw withdrawal in the hot plate test in rats when complexed with magnesium; has been used as a building block in the synthesis of GABA peptides with antinociceptive activity in rats; plasma levels of D- and L-pyroglutamic acid are increased in patients with end-stage renal disease
Catalog No. 50-291-2053 Supplier Cayman Chemical Supplier No. 36375500 G
A cyclized form of glutamic acid; decreases immobility time in the forced swim test and increases the latency to paw withdrawal in the hot plate test in rats when complexed with magnesium; has been used as a building block in the synthesis of GABA peptides with antinociceptive activity in rats; plasma levels of D- and L-pyroglutamic acid are increased in patients with end-stage renal disease