Learn More
Medchemexpress LLC DL-Pyroglutamic acid-d5 | 352431-30-2 | 99.21% | 134.14 | 5 MG
SDP
Catalog No. 5000454971
Encompass_Preferred
DL-Pyroglutamic acid-d5 is the deuterium labeled DL-Pyroglutamic acid. It is intended for research use only and can be utilized as a tracer and as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily for quantitation during drug development. Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
  • Intended for research use only
Catalog No. 50-004-54971 Supplier Medchemexpress LLC Supplier No. HYW012738S5MG
DL-Pyroglutamic acid-d5 is the deuterium labeled DL-Pyroglutamic acid. It is intended for research use only and can be utilized as a tracer and as an internal standard for quantitative analysis through methods such as NMR, GC-MS, or LC-MS. Stable heavy isotopes of hydrogen, carbon, and other elements are incorporated into drug molecules primarily for quantitation during drug development. Deuteration has shown potential to influence the pharmacokinetic and metabolic profiles of drugs.
  • Used as a tracer
  • Functions as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeling can affect pharmacokinetic and metabolic profiles of drugs
  • Intended for research use only