Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Pyroglutamic Acid, MP Biomedicals™

Catalog No. ICN10278025 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10278025 25 g
ICN10278080 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN10278025 Supplier MP Biomedicals, Inc Supplier No. 0210278025
Add to Cart
Add to Cart

Chemical Identifiers

CAS 149-87-1
Molecular Formula C5H7NO3
Molecular Weight (g/mol) 129.12
MDL Number MFCD00064322
InChI Key ODHCTXKNWHHXJC-UHFFFAOYNA-N
Synonym dl-pyroglutamic acid, 2-pyrrolidone-5-carboxylic acid, dl-proline, 5-oxo, 5-oxo-dl-proline, pyroglutamate, h-dl-pyr-oh, pyrrolidonecarboxylic acid, 5-oxoprolinate, d-+-pyroglutamic acid, dl-pidolic acid
PubChem CID 499
ChEBI CHEBI:16010
IUPAC Name 5-oxopyrrolidine-2-carboxylic acid
SMILES OC(=O)C1CCC(=O)N1

Specifications

Quantity 25 g
Formula Weight 129.1
Physical Form Solid
Chemical Name or Material DL-Pyroglutamic Acid
EINECSNumber 205-748-3
Recommended Storage Store at room temperature.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.