Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-Serine methyl ester hydrochloride, 99%

Catalog No. AC207451000
Change view
Click to view available options
Quantity:
10 g
100 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC207451000 100 g Glass Bottle
AC207450100 10 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC207451000 Supplier Thermo Scientific Chemicals Supplier No. 207451000
Add to Cart

missing translation for 'validMainframeMsg'

C4H9NO3·HCl

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C4H9NO3·HCl
MDL Number MFCD00012593

Specifications

Color White
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass Bottle
Linear Formula HOCH2CH(NH2)CO2CH3·HCl
Beilstein 04, 512
Quantity 100 g
Formula Weight 155.58
Percent Purity ≥98.5%
Physical Form Crystalline Powder
Chemical Name or Material DL-Serine methyleester hydrochloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.