Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-SulForaphane, 97%, MP Biomedicals™

Catalog No. ICN19378125 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN19378125 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. ICN19378125 Supplier MP Biomedicals, Inc Supplier No. 0219378125
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 4478-93-7
Molecular Formula C6H11NOS2
Molecular Weight (g/mol) 177.28
MDL Number MFCD00198068,MFCD09039283
InChI Key SUVMJBTUFCVSAD-UHFFFAOYNA-N
Synonym sulforaphane, dl-sulforaphane, sulforafan, 1-isothiocyanato-4-methylsulfinyl butane, d,l-sulforaphane, sulforaphane unspecified, r,s-sulforaphane, ccris 7221, 1-isothiocyanato-4-methylsulfinyl-butane, butane, 1-isothiocyanato-4-methylsulfinyl
PubChem CID 5350
ChEBI CHEBI:47807
IUPAC Name 1-isothiocyanato-4-methanesulfinylbutane
SMILES CS(=O)CCCCN=C=S

Specifications

Color Yellow
Assay Percent Range 0.97
Quantity 25 mg
Formula Weight 177.3
Physical Form Liquid
Chemical Name or Material DL-Sulforaphane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.