Learn More
Medchemexpress LLC DL-sulforaphane glutathione | 289711-21-3 | 484.61 g·mol⁻¹ | C16H28N4O7S3 | 5 MG
SDP
Catalog No. 5000285001
Encompass_Preferred
DL-Sulforaphane glutathione (DL-SFN-GSH) is the glutathione conjugate metabolite of sulforaphane, reported in rat plasma. It is supplied for research use and commonly used as an analytical reference in metabolic and pharmacokinetic studies to investigate sulforaphane biotransformation.
  • Metabolite of sulforaphane identified in vivo.
  • Suitable as an analytical standard for metabolic and PK studies.
  • Provided in a small research pack for analytical applications.
  • Chemical formula C16H28N4O7S3; molecular weight ~484.61 g·mol⁻¹.
  • For research use only; not intended for human or clinical use.
Catalog No. 50-002-85001 Supplier Medchemexpress LLC Supplier No. HYW5882485MG
DL-Sulforaphane glutathione (DL-SFN-GSH) is the glutathione conjugate metabolite of sulforaphane, reported in rat plasma. It is supplied for research use and commonly used as an analytical reference in metabolic and pharmacokinetic studies to investigate sulforaphane biotransformation.
  • Metabolite of sulforaphane identified in vivo.
  • Suitable as an analytical standard for metabolic and PK studies.
  • Provided in a small research pack for analytical applications.
  • Chemical formula C16H28N4O7S3; molecular weight ~484.61 g·mol⁻¹.
  • For research use only; not intended for human or clinical use.