Learn More
Medchemexpress LLC Dl-sulforaphane n-acetyl-l-cysteine | 334829-66-2 | 99.9% | 340.48 g/mol | C11H20N2O4S3 | 5 MG
SDP
Catalog No. 5000262126
Encompass_Preferred
DL-Sulforaphane N-acetyl-L-cysteine is a sulforaphane metabolite used in research as an orally active histone deacetylase (HDAC) inhibitor. It is reported to have an extended half-life and improved blood-brain barrier permeability versus the parent compound, and has been used to study autophagy, HDAC activity, and anticancer mechanisms. Supplied as a high-purity solid for laboratory research.
  • Orally active HDAC inhibitor and sulforaphane metabolite.
  • Longer half-life and improved blood-brain barrier permeability compared with parent compound.
  • Reported to induce autophagy, cell-cycle arrest, and apoptosis in certain cancer cell lines.
  • High-purity solid suitable for research applications.
  • Soluble in DMSO; store powder frozen for long-term stability.
Catalog No. 50-002-62126 Supplier Medchemexpress LLC Supplier No. HY1322425MG
DL-Sulforaphane N-acetyl-L-cysteine is a sulforaphane metabolite used in research as an orally active histone deacetylase (HDAC) inhibitor. It is reported to have an extended half-life and improved blood-brain barrier permeability versus the parent compound, and has been used to study autophagy, HDAC activity, and anticancer mechanisms. Supplied as a high-purity solid for laboratory research.
  • Orally active HDAC inhibitor and sulforaphane metabolite.
  • Longer half-life and improved blood-brain barrier permeability compared with parent compound.
  • Reported to induce autophagy, cell-cycle arrest, and apoptosis in certain cancer cell lines.
  • High-purity solid suitable for research applications.
  • Soluble in DMSO; store powder frozen for long-term stability.