Learn More
Cayman Chemical DL-Sulforaphn GlutathIon 5mg
SDP
Catalog No. 502910751
Encompass_Preferred
A metabolite of sulforaphane; increases mRNA expression of GSTA1 and UGT1A1 in HepG2 cells at 15 µM
Catalog No. 50-291-0751 Supplier Cayman Chemical Supplier No. 347315 MG
A metabolite of sulforaphane; increases mRNA expression of GSTA1 and UGT1A1 in HepG2 cells at 15 µM