Learn More
Medchemexpress LLC DL-tartaric acid | 133-37-9 | 98.0% | 150.09 g/mol | C4H6O6 | 5g
SDP
Catalog No. 5000266597
Encompass_Preferred
DL-Tartaric acid is an orally active and non-racemic mixture of L- and D-tartaric acid with antioxidant activity 14C-labeled DL-Tartaric acid is nephrotoxic[1][2][3]
Catalog No. 50-002-66597 Supplier Medchemexpress LLC Supplier No. HYY13155G
DL-Tartaric acid is an orally active and non-racemic mixture of L- and D-tartaric acid with antioxidant activity 14C-labeled DL-Tartaric acid is nephrotoxic[1][2][3]