Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Terebic Acid 98+%, Thermo Scientific™

Catalog No. AC198890100
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC198890100 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC198890100 Supplier Thermo Scientific Chemicals Supplier No. 198890100

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79-91-4
Molecular Formula C7H10O4
Molecular Weight (g/mol) 158.153
InChI Key UZBOWOQARWWIER-UHFFFAOYSA-N
Synonym terebic acid, tetrahydro-2,2-dimethyl-5-oxo-3-furoic acid, 3-furancarboxylic acid, tetrahydro-2,2-dimethyl-5-oxo, 2,2-dimethyl-5-oxotetrahydrofuran-3-carboxylic acid, terebinic acid, 2,2-dimethyl-5-oxo-2,3,4-trihydrofuran-3-carboxylic acid, terebinsaure, 2,2-dimethyl-5-oxo-tetrahydro-furan-3-carboxylic acid, 2,2-dimethyl-5-oxotetrahydro-3-furancarboxylic acid #, 3-furancarboxylic acid,tetrahydro-2,2-dimethyl-5-oxo
PubChem CID 101540
IUPAC Name 2,2-dimethyl-5-oxooxolane-3-carboxylic acid
SMILES CC1(C(CC(=O)O1)C(=O)O)C

Specifications

Melting Point 175°C to 180°C
Color White
Infrared Spectrum Authentic
Quantity 10 g
Merck Index 15, 9304
Formula Weight 158.15
Percent Purity ≥98.0%
Physical Form Fine Crystalline Powder
Chemical Name or Material DL-Terebic acid, 98+%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.