Learn More
Research Products International Corp DL-Thioctic Acid, 1 G

Supplier: Research Products International Corp T186001.0
Substance Name: DL-a-Lipoic acid
CAS Number: 1077-28-7
Molecular Formula: C8H14O2S2
Molecular Weight: 206.3
Chemical Purity: >99%
Soluble in: Ethanol
Store at: Room Temperature
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.