Learn More
Abcam D,L-Thioctic acid (DL-alpha-Lipoic acid), Non-specific free radical scavenger, 25G
Catalog No. 502706146
Encompass_Preferred
MW 206.3 Da, Purity >98%. Non-specific free radical scavenger. Prevents lipid peroxidation and promotes mitochondrial permeability transition in vitro. Anti-oxidant. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-6146 Supplier Abcam Supplier No. AB14295225G
MW 206.3 Da, Purity >98%. Non-specific free radical scavenger. Prevents lipid peroxidation and promotes mitochondrial permeability transition in vitro. Anti-oxidant. Active in vivo and in vitro.
The product is subject to the following: Abcam Restricted Use Statement