Learn More
Alkali Scientific DL Thioctic Acid [DL a Lipoic acid], 1 G
SDP
Catalog No. 5000660394
Encompass_Preferred
DL Thioctic Acid (DL Alpha Lipoic Acid), 1G, is a potent antioxidant used in various biological and chemical applications. This formulation is a racemic mixture of both the D- and L- forms of alpha-lipoic acid, a sulfur-containing fatty acid that plays a crucial role in cellular metabolism and acts as a cofactor for several mitochondrial enzymes. In addition to its antioxidant properties, DL Alpha Lipoic Acid has shown therapeutic potential in neuroprotective and anti-inflammatory treatments. It is commonly used in laboratory research for cell protection studies and metabolic assays and is also a key ingredient in pharmaceutical formulations for supporting liver health and treating diabetic neuropathy.
Catalog No. 50-006-60394 Supplier Alkali Scientific Supplier No. DLT001CP
DL Thioctic Acid (DL Alpha Lipoic Acid), 1G, is a potent antioxidant used in various biological and chemical applications. This formulation is a racemic mixture of both the D- and L- forms of alpha-lipoic acid, a sulfur-containing fatty acid that plays a crucial role in cellular metabolism and acts as a cofactor for several mitochondrial enzymes. In addition to its antioxidant properties, DL Alpha Lipoic Acid has shown therapeutic potential in neuroprotective and anti-inflammatory treatments. It is commonly used in laboratory research for cell protection studies and metabolic assays and is also a key ingredient in pharmaceutical formulations for supporting liver health and treating diabetic neuropathy.