Learn More
Alkali Scientific DL Thioctic Acid [DL a Lipoic acid], 25 G
SDP
Catalog No. 5000661582
Encompass_Preferred
DL Thioctic Acid (DL Alpha Lipoic Acid), 25G, is a powerful antioxidant and coenzyme involved in mitochondrial metabolism. This formulation is available in a 25g bulk package, ideal for laboratory research, pharmaceutical applications, and clinical studies. The racemic mixture of both D- and L-alpha-lipoic acid plays an essential role in the mitochondrial energy production process, offering protection against oxidative stress and free radicals. It is widely used in research to study its neuroprotective properties and its ability to improve insulin sensitivity, enhance vascular health, and support liver function. Its antioxidant properties also make it a key ingredient in various therapeutic and cosmetic formulations.
Catalog No. 50-006-61582 Supplier Alkali Scientific Supplier No. DLT002CP
DL Thioctic Acid (DL Alpha Lipoic Acid), 25G, is a powerful antioxidant and coenzyme involved in mitochondrial metabolism. This formulation is available in a 25g bulk package, ideal for laboratory research, pharmaceutical applications, and clinical studies. The racemic mixture of both D- and L-alpha-lipoic acid plays an essential role in the mitochondrial energy production process, offering protection against oxidative stress and free radicals. It is widely used in research to study its neuroprotective properties and its ability to improve insulin sensitivity, enhance vascular health, and support liver function. Its antioxidant properties also make it a key ingredient in various therapeutic and cosmetic formulations.