Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Thioctic Acid, Fisher BioReagents

Catalog No. BP26825 Shop All Fisherbrand Products
Change view
Click to view available options
Amber Glass; 5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
BP26825 Amber Glass; 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BP26825

missing translation for 'validMainframeMsg'

DL-Thioctic Acid is a growth factor for many bacteria and protozoa; prosthetic group, coenzyme, or substrate in plants, microorganisms, and animal tissue.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1077-28-7
Melting Point 60°C
Color Yellow
Boiling Point 160°C
Assay Percent Range ≥99.0 %
Packaging Amber Glass
Molecular Formula C8H14O2S2
MDL Number MFCD00005474
Quantity 5 g
Merck Index 15, 9479
Synonym dl-thioctic acid, thioctic acid, alpha-lipoic acid, 5-1,2-dithiolan-3-yl pentanoic acid, dl-alpha-lipoic acid, 1,2-dithiolane-3-pentanoic acid, 6,8-thioctic acid, thioctacid, 6-thioctic acid, alpha lipoic acid
InChI Key AGBQKNBQESQNJD-UHFFFAOYNA-N
SMILES OC(=O)CCCCC1CCSS1
IUPAC Name 5-(1,2-dithiolan-3-yl)pentanoic acid
Molecular Weight (g/mol) 206.32
PubChem CID 864
ChEBI CHEBI:16494
Formula Weight 206.32
Percent Purity 99.0% min.
Physical Form Powder or Solid
Chemical Name or Material DL-Thioctic Acid
Show More Show Less
Safety handling Check Alert storage code Red
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.