Learn More
Apexbio Technology LLC DL-threo-2-methylisocitrate sodium 1mg
SDP
Catalog No. 5000088702
Encompass_Preferred
DL-threo-2-methylisocitrate sodium is a substrate utilized by isocitrate lyase 1 (ICL1) It is designed to facilitate the investigation of ICL1 activity and substrate specificity thereby elucidating the enzyme s roles in the glyoxylate and methylcitrate cycles DL-threo-2-methylisocitrate sodium exerts its biological utility primarily by serving as a substrate for ICL1-mediated cleavage reactions Compared to threo-isocitrate ICL1 displays distinct enzymatic kinetic parameters toward this compound Based on these properties DL-threo-2-methylisocitrate sodium holds research potential for studying microbial metabolic pathways involving ICL1
Catalog No. 50-000-88702 Supplier Apexbio Technology LLC Supplier No. B13101
DL-threo-2-methylisocitrate sodium is a substrate utilized by isocitrate lyase 1 (ICL1) It is designed to facilitate the investigation of ICL1 activity and substrate specificity thereby elucidating the enzyme s roles in the glyoxylate and methylcitrate cycles DL-threo-2-methylisocitrate sodium exerts its biological utility primarily by serving as a substrate for ICL1-mediated cleavage reactions Compared to threo-isocitrate ICL1 displays distinct enzymatic kinetic parameters toward this compound Based on these properties DL-threo-2-methylisocitrate sodium holds research potential for studying microbial metabolic pathways involving ICL1