Learn More
Medchemexpress LLC DL-threo-3-hydroxyaspartic acid | 4294-45-5 | MFCD00036749 | 99.8% | 149.10 g/mol | C4H7NO5 | 25 MG
SDP
Catalog No. 5000213269
Encompass_Preferred
DL-threo-3-hydroxyaspartic acid is a research reagent that acts as a glutamate uptake inhibitor to block glutamate transport in biochemical and physiological studies. It is supplied as a white to off-white solid for laboratory use and is intended for research purposes only.
  • Glutamate uptake inhibitor for research applications.
  • High purity suitable for biochemical and pharmacological assays.
  • Supplied in small milligram pack sizes for preclinical work.
  • Recommended cold storage for long-term stability.
  • White to off-white solid, compatible with common solvents for dissolution.
Catalog No. 50-002-13269 Supplier Medchemexpress LLC Supplier No. HY100801A25MG
DL-threo-3-hydroxyaspartic acid is a research reagent that acts as a glutamate uptake inhibitor to block glutamate transport in biochemical and physiological studies. It is supplied as a white to off-white solid for laboratory use and is intended for research purposes only.
  • Glutamate uptake inhibitor for research applications.
  • High purity suitable for biochemical and pharmacological assays.
  • Supplied in small milligram pack sizes for preclinical work.
  • Recommended cold storage for long-term stability.
  • White to off-white solid, compatible with common solvents for dissolution.