Learn More
Medchemexpress LLC DL-threo-Chloramphenicol-d5 | 1420043-66-8 | 99.2% | 5 MG
SDP
Catalog No. 5000406916
Encompass_Preferred
DL-threo-Chloramphenicol-d5 is a deuterium labeled DL-threo-Chloramphenicol. DL-threo-Chloramphenicol is the racemate of Chloramphenicol. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-06916 Supplier Medchemexpress LLC Supplier No. HYB0239S15MG
DL-threo-Chloramphenicol-d5 is a deuterium labeled DL-threo-Chloramphenicol. DL-threo-Chloramphenicol is the racemate of Chloramphenicol. This product is for research use only.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.