Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DL-threo-PDMP hydroc 10mg | 80938-69-8 | 10 MG
SDP

Catalog No. 5000227160
Encompass_Preferred

DL-threo-PDMP hydrochloride is the hydrochloride salt of DL-threo-PDMP, a competitive glucosylceramide synthase (GCS) inhibitor used in biochemical and cellular research to modulate glycosphingolipid synthesis and study sphingolipid-related pathways.

  • High purity: 99.4% for reproducible results.
  • White to off-white solid suitable for analytical and preparative work.
  • Molecular weight: 427.02 and formula C23H39ClN2O3.
  • Soluble in DMSO up to 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at -20°C under nitrogen; in solution, -80°C for up to 6 months.
  • Available in small research pack sizes including 10 mg.

Catalog No. 50-002-27160 Supplier Medchemexpress LLC Supplier No. HY116392B10MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DL-threo-PDMP hydrochloride is the hydrochloride salt of DL-threo-PDMP, a competitive glucosylceramide synthase (GCS) inhibitor used in biochemical and cellular research to modulate glycosphingolipid synthesis and study sphingolipid-related pathways.

  • High purity: 99.4% for reproducible results.
  • White to off-white solid suitable for analytical and preparative work.
  • Molecular weight: 427.02 and formula C23H39ClN2O3.
  • Soluble in DMSO up to 100 mg/mL; ultrasonic assistance recommended.
  • Storage: store at -20°C under nitrogen; in solution, -80°C for up to 6 months.
  • Available in small research pack sizes including 10 mg.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.