Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DL-Thyronine 98.0+%, TCI America™
SDP

Catalog No. T0241100MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T0241100MG 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. T0241100MG Supplier TCI America Supplier No. T0241100MG
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 1034-10-2
Molecular Formula C15H15NO4
Molecular Weight (g/mol) 273.288
MDL Number MFCD00045864
InChI Key KKCIOUWDFWQUBT-UHFFFAOYSA-N
PubChem CID 102544
ChEBI CHEBI:30661
IUPAC Name 2-amino-3-[4-(4-hydroxyphenoxy)phenyl]propanoic acid
SMILES C1=CC(=CC=C1CC(C(=O)O)N)OC2=CC=C(C=C2)O

Specifications

Color White-Yellow
Quantity 100 mg
Formula Weight 273.29
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material DL-Thyronine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.