Learn More
Medchemexpress LLC DL-α-Tocopherol acetate | 52225-20-4 | 99.5% | 10 MM 1 ML
SDP
Catalog No. 5000412938
Encompass_Preferred
DL-α-Tocopherol acetate is a vitamin E derivative often included in the formulations of enteral nutrition.
  • Increases tissue cholesterol levels in liver, brain, testis, and heart.
  • Brain cholesterol levels are significantly higher than in other tissues when adjusted by protein level.
  • Increases the level of lipid peroxidation in liver, brain, and testis of mice.
Catalog No. 50-004-12938 Supplier Medchemexpress LLC Supplier No. HYB1278A10MM/1ML
DL-α-Tocopherol acetate is a vitamin E derivative often included in the formulations of enteral nutrition.
  • Increases tissue cholesterol levels in liver, brain, testis, and heart.
  • Brain cholesterol levels are significantly higher than in other tissues when adjusted by protein level.
  • Increases the level of lipid peroxidation in liver, brain, and testis of mice.