Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific Chemicals DL-alpha-Tocopheryl acetate, 98%

Catalog No. AC421070050
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC421070050 5 g Glass bottle
AC421070010 1 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC421070050 Supplier Thermo Scientific Chemicals Supplier No. 421070050
  • $195.00
    $151.65 / Each of 1

    Save $43.35

    22% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
View all promotions
Add to Cart

missing translation for 'validMainframeMsg'

CAS: 7695-91-2 | C31H52O3 | 472.75 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7695-91-2
Molecular Formula C31H52O3
Molecular Weight (g/mol) 472.75
MDL Number MFCD00072042
InChI Key ZAKOWWREFLAJOT-CEFNRUSXSA-N
Synonym vitamin e acetate, tocopherol acetate, alpha-tocopherol acetate, alfacol, ecofrol, d-alpha-tocopherol acetate, contopheron, tofaxin, ephynal acetate, econ
PubChem CID 86472
ChEBI CHEBI:32321
IUPAC Name [(2R)-2,5,7,8-tetramethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-yl] acetate
SMILES CC1=C2C(=C(C(=C1C)OC(=O)C)C)CCC(O2)(C)CCCC(C)CCCC(C)CCCC(C)C

Specifications

Color Green to Yellow
Flash Point 257°C
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 9653
Solubility Information Solubility in water: slightly soluble
Formula Weight 472.75
Percent Purity 98%
Physical Form Viscous Liquid
Chemical Name or Material DL-α-Tocopheryl acetate
Show More Show Less
EINECSNumber 231-710-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.