Learn More
Medchemexpress LLC DL-Tryptophan-d3 | 340257-61-6 | 99.29% | 207.24 | 1 MG
SDP
Catalog No. 5000452372
Encompass_Preferred
DL-Tryptophan-d3 is a deuterium labeled DL-Tryptophan, an endogenous metabolite. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52372 Supplier Medchemexpress LLC Supplier No. HYW012480S1MG
DL-Tryptophan-d3 is a deuterium labeled DL-Tryptophan, an endogenous metabolite. It is for research use only.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS