Learn More
Medchemexpress LLC DL-Tryptophan-d3 | 340257-61-6 | 99.3% | 207.24 | 10 MG
SDP
Catalog No. 5000452968
Encompass_Preferred
DL-Tryptophan-d3 is the deuterium labeled DL-Tryptophan. DL-Tryptophan is an endogenous metabolite. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
Catalog No. 50-004-52968 Supplier Medchemexpress LLC Supplier No. HYW012480S10MG
DL-Tryptophan-d3 is the deuterium labeled DL-Tryptophan. DL-Tryptophan is an endogenous metabolite. This compound can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS