Learn More
Cayman Chemical DL-Tryptophan octyl ester (hydrochloride), Purity: ≥98%, CAS: 6278-90-6, 1 g
SDP
Catalog No. 502967504
Encompass_Preferred
A stable, cell-permeable tryptophan derivative with an octyl ester protected COOH group
Catalog No. 50-296-7504 Supplier Cayman Chemical Supplier No. 175991 G
A stable, cell-permeable tryptophan derivative with an octyl ester protected COOH group