Learn More
Medchemexpress LLC DL-Valine-d8 | 203784-63-8 | 100.0% | 125.20 | 50 MG
SDP
Catalog No. 5000454812
Encompass_Preferred
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and have the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled
  • Intended for research use only
Catalog No. 50-004-54812 Supplier Medchemexpress LLC Supplier No. HYW012889S50MG
DL-Valine-d8 is the deuterium labeled DL-Valine. It can be used as a tracer and as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS. Stable heavy isotopes, like deuterium, are incorporated into drug molecules for quantitation during drug development and have the potential to affect pharmacokinetic and metabolic profiles of drugs.
  • Tracer
  • Internal standard for quantitative analysis by NMR, GC-MS, or LC-MS
  • Deuterium labeled
  • Intended for research use only