Learn More
Cayman Chemical DLaDIfluoromethylornIthIn 50mg
SDP
Catalog No. 502967028
Encompass_Preferred
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects
Catalog No. 50-296-7028 Supplier Cayman Chemical Supplier No. 1688950 MG
An irreversible inhibitor of ornithine decarboxylase that suppresses polyamine biosynthesis; displays antiangiogenic, cytostatic, and antiparasitic effects