Learn More
Aviva Systems Biology Corp DLAT Antibody - C-terminal region (ARP41995_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP41995_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLAT. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DLAT. Alternate names: DLTA, PDC-E2, PDCE2ApplicationWB. Species reactivity: Dog, Human, Mouse, Pig, Rat, Zebrafish
Catalog No. 50-014-58074