Learn More
Aviva Systems Biology Corp DLC1 Antibody - C-terminal region (ARP52138_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP52138_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLC1. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the C terminal region of human DLC1. Alternate names: ARHGAP7, FLJ21120, HP, STARD12, p122-RhoGAPApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-60686