Learn More
Enzo Life Sciences DLC1 monoclonal antibody (10D6) (100µg)
SDP
Supplier:  Enzo Life Sciences ALX804340C100
Encompass_Preferred
Clone: 10D6. Isotype: IgM. Immunogen: Recombinant mouse DLC1 (dynein light chain 1; LC8; PIN). Species reactivity: Human, Mouse. Applications: Flow Cytometry, WB. Purity: Affinity purified. Formulation: Liquid. In PBS containing 10mM sodium azide.
Catalog No. 50-200-9871