Learn More
Cayman Chemical DL-Cystathionine (hydrochloride), Purity: ≥95%, 100 mg
SDP
Catalog No. 502966336
Encompass_Preferred
An intermediate in the synthesis of cysteine that is produced from homocysteine and serine by the action of cystathionine-β-synthase; excess cystathionine in the urine is a hallmark of cystathioninuria
Catalog No. 50-296-6336 Supplier Cayman Chemical Supplier No. 17819100 MG
An intermediate in the synthesis of cysteine that is produced from homocysteine and serine by the action of cystathionine-β-synthase; excess cystathionine in the urine is a hallmark of cystathioninuria