Learn More
Aviva Systems Biology Corp DLD Antibody - middle region (ARP58455_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP58455_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLD. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human DLD. Alternate names: DLDH, E3, GCSL, LAD, PHE3ApplicationIHC, WB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-62312