Learn More
Eurofins Discoverx cAMP Hunter DLD1 HTR7D Gs Cell Line, 2 vials. (95-0163C13)
Supplier:  Eurofins Discoverx 950163C13
Encompass
The cAMP Hunter HTR7D Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. serotonin/5-HT) based activation of HTR7D GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type HTR7D, a naturally Gs coupled GPCR. Upon activation of HTR7D, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3916
Add to Cart
Request a Quote