Learn More
Eurofins Discoverx PathHunter DLD1 IL7R/CRLF2 Dimerization Cell Line, 2 vials. (93-1019C13)
Catalog No. 502001861
Encompass
The PathHunter IL7R/CRLF2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1861 Supplier Eurofins Discoverx Supplier No. 931019C13
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL7R/CRLF2 Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. TSLP) induced dimerization of IL7R/CRLF2 receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside CRLF2 target protein fused to EA. Upon ligand-induced dimerization of IL7R/CRLF2, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.