Learn More
Eurofins Discoverx PathHunter DLD1 IL7R/IL2RG Dimerization Cell Line, 2 vials. (93-0997C13)
Catalog No. 502001780
Encompass
The PathHunter IL7R/IL2RG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.
Catalog No. 50-200-1780 Supplier Eurofins Discoverx Supplier No. 930997C13
Add to Cart
Add to Cart
Request a Quote
The PathHunter IL7R/IL2RG Dimerization cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting receptor-receptor interaction. This cell line enables assessment of ligand (e.g. IL-7) induced dimerization of IL7R/IL2RG receptors. This product uses EFC technology. The cell line included overexpresses IL7R target protein fused to PK alongside IL2RG target protein fused to EA. Upon ligand-induced dimerization of IL7R/IL2RG, detectable EFC signal is produced. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/interleukins.