Learn More
Eurofins Discoverx cAMP Hunter DLD1 PTGER2 Gs Cell Line, 2 vials. (95-0176C13)
Catalog No. 502003966
Encompass
The cAMP Hunter PTGER2 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of PTGER2 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGER2, a naturally Gs coupled GPCR. Upon activation of PTGER2, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.
Catalog No. 50-200-3966 Supplier Eurofins Discoverx Supplier No. 950176C13
Add to Cart
Add to Cart
Request a Quote
The cAMP Hunter PTGER2 Gs cell line is a stable, engineered cell line for use in studying drug candidates (small molecules or biologics) targeting GPCRs. This cell line enables assessment of ligand (e.g. prostaglandin D2) based activation of PTGER2 GPCR activity via detection of increasing levels of intracellular cAMP. This product uses EFC technology. The cells overexpress wild type PTGER2, a naturally Gs coupled GPCR. Upon activation of PTGER2, the increase in levels of intracellular cAMP leads to an increased signal from EFC when the cAMP detection kit is applied. This stable modified cell line is provided as two vials of cryopreserved cells. For detailed product information, User Manual and datasheet, visit www.discoverx.com/gpcr.