Learn More
Cayman Chemical DLerythroPPMPhydrochlorId 10mg
SDP
Catalog No. 502932544
Encompass_Preferred
A stereoisomer of DL-threo-PPMP; has no effect on glucosylceramide synthetase activity; used as a negative control for DL-threo-PPMP, D-threo-PPMP, and L-threo-PPMP inhibitory activity
Catalog No. 50-293-2544 Supplier Cayman Chemical Supplier No. 2452410 MG
A stereoisomer of DL-threo-PPMP; has no effect on glucosylceramide synthetase activity; used as a negative control for DL-threo-PPMP, D-threo-PPMP, and L-threo-PPMP inhibitory activity