Learn More
Cayman Chemical DL-Folinic Acid (calcium salt), Purity: ≥98%, CAS: 1492-18-8, 100 mg
SDP
Catalog No. 502984364
Encompass_Preferred
A formylated form of tetrahydrofolic acid, the reduced form of folic acid; used to prevent toxicity following high-dose methotrexate therapy, to enhance the antitumor activity of 5-fluorouracil, and as a folate supplement
Catalog No. 50-298-4364 Supplier Cayman Chemical Supplier No. 20383100 MG
A formylated form of tetrahydrofolic acid, the reduced form of folic acid; used to prevent toxicity following high-dose methotrexate therapy, to enhance the antitumor activity of 5-fluorouracil, and as a folate supplement