Learn More
Aviva Systems Biology Corp DLG3 Antibody - N-terminal region (ARP43694_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP43694_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLG3. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DLG3. Alternate names: KIAA1232, MRX, MRX90, NE-Dlg, NEDLG, SAP102, XLMRApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat, Zebrafish
Catalog No. 50-014-58609