Learn More
Aviva Systems Biology Corp DLG7 Antibody - N-terminal region (ARP46205_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP46205_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against DLG7. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the N terminal region of human DLG7. Alternate names: DLG1, HURP, KIAA0008, DLG7ApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-014-59267