Learn More
Cayman Chemical DL-Glyceraldehyde-3-phosphate, CAS: 591-59-3, 5 mg
SDP
Catalog No. 502965318
Encompass_Preferred
An intermediate in several metabolic pathways, including glycolysis and gluconeogenesis
Catalog No. 50-296-5318 Supplier Cayman Chemical Supplier No. 178655 MG
An intermediate in several metabolic pathways, including glycolysis and gluconeogenesis