Learn More
Cayman Chemical DL-Glyceraldehyde-3-phosphate, CAS: 591-59-3, 10 mg
SDP
Catalog No. 502965357
Encompass_Preferred
An intermediate in several metabolic pathways, including glycolysis and gluconeogenesis
Catalog No. 50-296-5357 Supplier Cayman Chemical Supplier No. 1786510 MG
An intermediate in several metabolic pathways, including glycolysis and gluconeogenesis