Learn More
Medchemexpress LLC DLin-K-C3-DMA | 1217306-46-1 | 98.59% | C44H81NO2 | 100 MG
SDP
Catalog No. 5000354730
Encompass_Preferred
DLin-K-C3-DMA is a cationic lipid that can be used in the synthesis of nucleic acid-lipid particles for the delivery of nucleic acid.
  • Cationic lipid
  • Used in nucleic acid-lipid particle synthesis
  • For nucleic acid delivery
Catalog No. 50-003-54730 Supplier Medchemexpress LLC Supplier No. HY145225100MG
DLin-K-C3-DMA is a cationic lipid that can be used in the synthesis of nucleic acid-lipid particles for the delivery of nucleic acid.
  • Cationic lipid
  • Used in nucleic acid-lipid particle synthesis
  • For nucleic acid delivery