Learn More
Medchemexpress LLC DLin-K-C3-DMA | 1217306-46-1 | 98.59% | C44H81NO2 | 25 MG
SDP
Catalog No. 5000357372
Encompass_Preferred
DLin-K-C3-DMA is a cationic lipid that can be used in the synthesis of nucleic acid-lipid particles to deliver nucleic acids. It is for research use only.
  • Cationic lipid for nucleic acid delivery.
  • Used in the synthesis of nucleic acid-lipid particles.
Catalog No. 50-003-57372 Supplier Medchemexpress LLC Supplier No. 25MGHY145225
DLin-K-C3-DMA is a cationic lipid that can be used in the synthesis of nucleic acid-lipid particles to deliver nucleic acids. It is for research use only.
  • Cationic lipid for nucleic acid delivery.
  • Used in the synthesis of nucleic acid-lipid particles.