Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC DLin-K-C3-DMA | 1217306-46-1 | 98.6% | C44H81NO2 | 1 ML
SDP

Catalog No. 5000354583
Encompass_Preferred

DLin-K-C3-DMA is a cationic lipid used in the synthesis of nucleic acid-lipid particles for the delivery of nucleic acids. It is intended for research use only.

  • Cationic lipid for nucleic acid delivery
  • Appearance: colorless to light yellow liquid
  • Molecular weight: 656.12
  • Supplied as a 10 mM solution

Catalog No. 50-003-54583 Supplier Medchemexpress LLC Supplier No. HY14522510MM/1ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

DLin-K-C3-DMA is a cationic lipid used in the synthesis of nucleic acid-lipid particles for the delivery of nucleic acids. It is intended for research use only.

  • Cationic lipid for nucleic acid delivery
  • Appearance: colorless to light yellow liquid
  • Molecular weight: 656.12
  • Supplied as a 10 mM solution

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.