Learn More
TARGETMOL CHEMICALS INC DLin-KC2-DMA 10MG
SDP
Catalog No. 502878628
Encompass_Preferred
Also available in 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. DLin-KC2-DMA is a cation that can be used for siRNA delivery and is also an ionizable lipid. DLinKC2-DMA can bind to LNP and enable siRNA-mediated GAPDH gene silencing. Purity 97.74%
Catalog No. 50-287-8628 Supplier TARGETMOL CHEMICALS INC Supplier No. T1513910MG
Also available in 2 mg, 5 mg, 25 mg, 50 mg, 100 mg, 200 mg and bulk. Please contact Fisher for quotes. DLin-KC2-DMA is a cation that can be used for siRNA delivery and is also an ionizable lipid. DLinKC2-DMA can bind to LNP and enable siRNA-mediated GAPDH gene silencing. Purity 97.74%